Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • NFkB p50 Antibodies

          OriGene

          NFKB1 Monoclonal Antibody (OTI3A9)

          View all (15) NFkB p50 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite NFKB1 Monoclonal Antibody (OTI3A9)

          • Antibody Testing Data (2)
          NFKB1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NFKB1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NFKB1 Antibody (TA813232) in WB

          HEK293T cells were transfected with the pCMV6-ENTRY control (Left lane) or pCMV6-ENTRY NFKB1 (RC208384, Right lane) cDNA for 48 hrs and lysed. Equivalent amounts of cell lysates (5 ug per lane) were separated by SDS-PAGE and immunoblotted with anti-NFKB1 (1:500). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NFKB1 Antibody in Western Blot (WB)
          NFKB1 Antibody in Western Blot (WB)

          Product Details

          TA813232

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:250-1:500
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          OTI3A9

          Immunogen

          Full length human recombinant protein of human NFKB1 (NP_003989) produced in HEK293T cell.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS with 50% glycerol, 1% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          NFkB is a nuclear transcription factor activated by various extra and intracellular stimuli such as cytokines, UV radiation, stress, in injury, and by bacterial and viral products. NFkB is involved in regulation of various cellular events including cell growth, differentiation, proliferation, apoptosis and inflammation. NFKB1 (p50) is a 50KDa functional sub-unit of NF-kB and a member of Rel protein family which is synthesized as a p105 precursor protein and consists of an N-terminal conserved RHD-region containing nuclear localization signal, DNA-binding and dimerization domains. NFKB1 (p50) forms homodimers or heterodimerizes with p65, forming the functional NFkB factor. Further, NFKB1 (p50) directs the nuclear translocation of NF-kB and is instrumental in its DNA-binding. The NFkB complex is expressed in most cell types, and is primarily found in the cytoplasm in an inactive state in association with I-kappa-B. Phosphorylation of I-kappa-B releases the complex, allowing translocation to the nucleus and modification of gene expression. Phosphorylation of the p50 subunit at the Ser-337 residue has been shown to significantly increase DNA binding efficiency of the NFkB protein complex. The pathological role of NFkB has been suggested in AIDS, hematogenic cancer cell metastasis and rheumatoid arthritis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DKFZp686C01211; DNA-binding factor KBF1; EBP-1; MGC54151; NF KB; NF-kappabeta; NF-kB p50; nuclear factor; nuclear factor kappa b; nuclear factor kappa-B DNA binding subunit; Nuclear factor NF-kappa-B p105 subunit; nuclear factor NF-kappa-B p50 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1; OTTHUMP00000219572

          View more View less

          Gene Aliases: CVID12; EBP-1; KBF1; NF-kappa-B; NF-kappaB; NF-kB1; NFkappaB; NFKB-p105; NFKB-p50; NFKB1; p105; p50

          View more View less

          UniProt ID: (Human) P19838

          View more View less

          Entrez Gene ID: (Human) 4790

          View more View less

          Function(s)
          regulatory region DNA binding transcription regulatory region sequence-specific DNA binding RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II distal enhancer sequence-specific DNA binding transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding transcription factor binding heat shock protein binding identical protein binding protein homodimerization activity actinin binding transcription regulatory region DNA binding protein heterodimerization activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter stimulatory C-type lectin receptor signaling pathway transcription from RNA polymerase II promoter apoptotic process inflammatory response response to oxidative stress I-kappaB kinase/NF-kappaB signaling positive regulation of macrophage derived foam cell differentiation positive regulation of lipid storage negative regulation of calcidiol 1-monooxygenase activity negative regulation of vitamin D biosynthetic process membrane protein intracellular domain proteolysis negative regulation of cellular protein metabolic process negative regulation of cholesterol transport positive regulation of type I interferon production response to muscle stretch NIK/NF-kappaB signaling Fc-epsilon receptor signaling pathway negative regulation of apoptotic process negative regulation of interleukin-12 biosynthetic process innate immune response positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter response to copper ion negative regulation of inflammatory response T cell receptor signaling pathway positive regulation of NF-kappaB transcription factor activity stress-activated MAPK cascade cellular response to lipopolysaccharide cellular response to mechanical stimulus cellular response to nicotine cellular response to carbohydrate stimulus cellular response to interleukin-1 cellular response to interleukin-6 cellular response to tumor necrosis factor cellular response to dsRNA cellular response to peptide hormone stimulus positive regulation of canonical Wnt signaling pathway positive regulation of hyaluronan biosynthetic process cellular response to diterpene cellular response to glucoside cellular response to brain-derived neurotrophic factor stimulus positive regulation of miRNA metabolic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0