Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • NFKBIL2 Antibodies

          Abnova

          NFKBIL2 Polyclonal Antibody, MaxPab™

          View all (9) NFKBIL2 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite NFKBIL2 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          NFKBIL2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NFKBIL2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NFKBIL2 Antibody (H00004796-B01P) in WB

          Western Blot analysis of NFKBIL2 expression in transfected 293T cell line by NFKBIL2 MaxPab polyclonal antibody. Lane 1: NFKBIL2 transfected lysate(134.09 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NFKBIL2 Antibody in Western Blot (WB)

          Product Details

          H00004796-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NFKBIL2 (AAH08782.1, 1 a.a. approximately 1219 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.76 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MRTRLYLNLG LTFESLQQTA LCNDYFRKSI FLAEQNHLYE DLFRARYNLG TIHWRAGQHS QAMRCLEGAR ECAHTMRKRF MESECCVVIA QVLQDLGDFL AAKRALKKAY RLGSQKPVQR AAICQNLQHV LAVVRLQQQL EEAEGRDPQG AMVICEQLGD LFSKAGDFPR AAEAYQKQLR FAELLDRPGA ERAIIHVSLA TTLGDMKDHH GAVRHYEEEL RLRSGNVLEE AKTWLNIALS REEAGDAYEL LAPCFQKALS CAQQAQRPQL QRQVLQHLHT VQLRLQPQEA PETETRLREL SVAEDEDEEE EAEEAAATAE SEALEAGEVE LSEGEDDTDG LTPQLEEDEE LQGHLGRRKG SKWNRRNDMG ETLLHRACIE GQLRRVQDLV RQGHPLNPRD YCGWTPLHEA CNYGHLEIVR FLLDHGAAVD DPGGQGCEGI TPLHDALNCG HFEVAELLLE RGASVTLRTR KGLSPLETLQ QWVKLYRRDL DLETRQKARA MEMLLQAAAS GQDPHSSQAF HTPSSLLFDP ETSPPLSPCP EPPSNSTRLP EASQVHVRVS PGQAAPAMAR PRRSRHGPAS SSSSSEGEDS AGPARPSQKR PRCSATAQRV AAWTPGPASN REAATASTSR AAYQAAIRGV GSAQSRLGPG PPRGHSKALA PQAALIPEEE CLAGDWLELD MPLTRSRRPR PRGTGDNRRP SSTSGSDSEE SRPRARAKQV RLTCMQSCSA PVNAGPSSLA SEPPGSPSTP RVSEPSGDSS AAGQPLGPAP PPPIRVRVQV QDHLFLIPVP HSSDTHSVAW LAEQAAQRYY QTCGLLPRLT LRKEGALLAP QDLIPDVLQS NDEVLAEVTS WDLPPLTDRY RRACQSLGQG EHQQVLQAVE LQGLGLSFSA CSLALDQAQL TPLLRALKLH TALRELRLAG NRLGDKCVAE LVAALGTMPS LALLDLSSNH LGPEGLRQLA MGLPGQATLQ SLEELDLSMN PLGDGCGQSL ASLLHACPLL STLRLQACGF GPSFFLSHQT ALGSAFQDAE HLKTLSLSYN ALGAPALART LQSLPAGTLL HLELSSVAAG KGDSDLMEPV FRYLAKEGCA LAHLTLSANH LGDKAVRDLC RCLSLCPSLI SLDLSANPEI SCASLEELLS TLQKRPQGLS FLGLSGCAVQ GPLGLGLWDK IAAQLRELQL CSRRLCAEDR DALRQLQPSR PGPGECTLDH GSKLFFRRL

          Target Information

          The protein encoded by this gene is thought to be a negative regulator of NF-kappa-B mediated transcription. The encoded protein may bind NF-kappa-B complexes and trap them in the cytoplasm, preventing them from entering the nucleus and interacting with the DNA. Phosphorylation of this protein targets it for degradation by the ubiquitination pathway, which frees the NF-kappa-B complexes to enter the nucleus.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DNA repair protein; I-kappa-B-related protein; ikappaBR; Inhibitor of kappa B-related protein; NF-kappa-B inhibitor-like protein 2; Nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 2; tonsoku-like; Tonsoku-like protein

          View more View less

          Gene Aliases: IKBR; NFKBIL2; TONSL

          View more View less

          UniProt ID: (Human) Q96HA7

          View more View less

          Entrez Gene ID: (Human) 4796

          View more View less

          Function(s)
          transcription corepressor activity protein binding histone binding
          Process(es)
          double-strand break repair via homologous recombination replication fork processing cytoplasmic sequestering of transcription factor negative regulation of nucleic acid-templated transcription
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0