Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AHLHSFSMM PSSACEAMVG HEMASDSSNT SLPFSNMGNP MNTTQLGKSL FQWQVEQEES KLANISQDQF LSKDADGDTF LHIAVAQGRR ALSYVLARKM NALHMLDIKE HNGQSAFQVA VAANQHLIVQ DLVNIGAQVN TTDCWGRTPL HVCAEKGHSQ VLQAIQKGAV GSNQFVDLEA TNYDGLTPLH CAVIAHNAVV HELQRNQQPH SPEVQELLLK NKSLVDTIKC LIQMGAAVEA KDRKSGRTAL HLAAEEANLE LIRLFLELPS CLSFVNAKAY NGNTALHVAA SLQYRLTQLD AVRLLMRKGA DPSTRNLENE QPVHLVPDGP VGEQIRRILK GKSIQQRAPP Y (369-718 aa encoded by BC060800)
I-kappa-B-zeta (also known as MAIL and INAP) is an ankyrin-repeat-containing nuclear protein that is highly homologous to the IkappaB family member Bcl-3. Transcription of IkappaBzeta is upregulated by stimulation with TLR ligands, IL-1 and IL-6 in cultured B-lymphocytes and monocytes/macrophages, but only faintly so in T-lymphocytes, fibroblasts, and endothelial cells. IkappaB-zeta preferentially associates with the NF-kappaB subunit p50 rather than p65 and recombinant IkappaB-zeta proteins inhibit the DNA binding of the p65/p50 heterodimer and the p50/p50 homodimer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ30225; FLJ34463; I-kappa-B-zeta; Ikappa B-zeta variant 3; IkappaB-zeta; ikappaBzeta; IkB-zeta; IL-1 inducible nuclear ankyrin-repeat protein; INAP; MAIL; Molecule possessing ankyrin repeats induced by lipopolysaccharide; molecule possessing ankyrin-repeats induced by lipopolysaccharide; NF-kappa-B inhibitor zeta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, zeta; OTTHUMP00000214340
Gene Aliases: AA408868; IKBZ; INAP; MAIL; NFKBIZ; RGD1310834
UniProt ID: (Human) Q9BYH8, (Mouse) Q9EST8
Entrez Gene ID: (Human) 64332, (Rat) 304005, (Mouse) 80859
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support