Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen Sequence: LGINEISSSF FSLLLEILLL ESQASLPMLE ERVLDWQSSP ASSLNSWFSA APNWAELVLP ALQYLAGESR AVPSSFSPFV EFKEKTQQWK LLGQSQDNEK ELAALFQLWL ETKDQAFCKQ ENEDSS
NFRKB (nuclear factor related to kappa-B-binding protein) was first identified as a factor that could bind a variant NF-kappa B site in the interleukin-2 receptor alpha (IL-2R alpha) gene. NFRKB appears to be a transcription factor central to immune cell development. Evidence of this comes from the identification of NFRKB as an amplified gene in several cases of acute myeloid leukemia. Additionally, in another clinical case, a partial deletion of the chromosomal region that includes NFRKB is shown to be associated with combined cellular immunodeficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-binding protein R kappa B; DNA-binding protein R kappa-B; INO80 complex subunit G; Nuclear factor related to kappa-B-binding protein
Gene Aliases: INO80G; NFRKB
UniProt ID: (Human) Q6P4R8
Entrez Gene ID: (Human) 4798
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support