Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEQYTANSN SSTEQIVVQA GQIQQQVQGQ PLMVQVSGGQ LITSTGQPIM VQAVPGGQGQ TIMQVPVSGT QGLQQIQLVP PGQIQIQGGQ AVQVQGQQGQ TQQIIIQQPQ TAVTAGQTQT QQQIAVQGQQ VAQTAEGQTI VYQPVNADGT ILQQVTVPVS GMITIPAASL AGAQIVQTGA NTNTTSSGQG TVTVTLPVAG NVVNSGGMVM MVPGAGSVPA IQRIPLPGAE MLEEEPLYVN AKQYNRILKR RQARAKLEAE GKIPKERRKY LHESRHRHAM ARKRGEGG (1-287 aa encoded by BC039244)
The SMN1 gene is part of a 500 kb inverted duplication on chromosome 5q13. This duplicated region contains at least four genes and repetitive elements which make it prone to rearrangements and deletions. The repetitiveness and complexity of the sequence have also caused difficulty in determining the organization of this genomic region. The telomeric and centromeric copies of this gene are nearly identical and encode the same protein - survival motor neuron protein. The SMN complex plays a catalyst role in the assemble of small nuclear ribonucleoproteins, the building blocks of the spliceosome. Mutations in the SMN1 gene are known to cause spinal muscular atrophy 1/2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAAT box DNA-binding protein subunit A; CAAT-box DNA binding protein subunit A; CAAT-box DNA-binding protein subunit A; CBF-A; CBF-B; CCAAT-binding transcription factor subunit A; CCAAT-binding transcription factor subunit B; FLJ11236; HAP2 CCAAT-binding protein; nf-y; NF-YA; Nuclear transcription factor Y subunit A; Nuclear transcription factor Y subunit alpha; nuclear transcription factor Y, alpha; Transcription factor NF-Y, A subunit
Gene Aliases: AA407810; CBF-A; CBF-B; HAP2; NF-YA; NFYA; SEZ-10; SEZ10
UniProt ID: (Human) P23511, (Mouse) P23708, (Rat) P18576
Entrez Gene ID: (Human) 4800, (Mouse) 18044, (Rat) 29508
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support