Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total NFYB.
The CCAAT-binding factor NF-Y is a heteromeric transcription factor that specifically binds to CCAAT sequences in many eukaryotic genes. NF-Y is made up of three subunits, NF-YA, NF-YB and NF-YC. All three components are necessary for DNA binding. In each NF-Y subunit, the segment needed for formation of the NF-Y-DNA complex is conserved from yeast to human. These conserved segments are homologous to the histone-fold motif of eukaryotic histones. The DNA-binding domains of the NF-YB and NF-YC subunits have been suggested to interact through a protein-protein, histone-fold "handshake" motif in a manner analogous to the histone proteins H2B and H2A, respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAAT box DNA-binding protein subunit B; CAAT-box DNA-binding protein subunit B; CBF-A; CBF-B; CCAAT binding transcription factor of CBF-B/NFY-B; CCAAT-binding transcription factor subunit A; CCAAT-binding transcription factor subunit B; nf-y; NF-YB; nuclear transcription factor - Y beta; Nuclear transcription factor Y subunit B; Nuclear transcription factor Y subunit beta; nuclear transcription factor Y, beta; Transcription factor NF-Y, B subunit
Gene Aliases: AA985999; CBF-A; CBF-B; CBFB; HAP3; NF-YB; Nfya; NFYB
UniProt ID: (Human) P25208, (Mouse) P63139, (Rat) P63140
Entrez Gene ID: (Human) 4801, (Mouse) 18045, (Rat) 25336
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support