Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MSRVLVPCHV KGSVALQVGD VRTSQGRPGV LVIDVTFPSV APFELQEITF KNYYTAFLSI RVRQYTSAHT PAKWVTCLRD YCLMPDPHSE EGAQEYVSLF KHQMLCDMAR ISELRLILRQ PSPLWLSFTV EELQIYQQGP KSPSVTFPKW LSHPVPCEQP ALLREGLPDP SRVSSEVQQM WALTEMIRAS HTSARIGRFD VDGCYDLNLL SYT
This protein encoded by this gene localizes to the nucleus and is expressed in numerous tissues including brain, testis, liver, and kidney. Computer predictions indicate that this region of the 3' UTR contains hairpin-forming self-complementary sequence which is possibly excised after transcription. This gene has a pseudogene on chromosome X.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nicolin-1; NPCEDRG; p24; PGs5; Tubulin polyglutamylase complex subunit 5
Gene Aliases: 1500032A17Rik; NICN1
UniProt ID: (Human) Q9BSH3, (Mouse) Q9CQM0
Entrez Gene ID: (Human) 84276, (Rat) 619581, (Mouse) 66257
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support