Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • NKG2A/NKG2C/NKG2E Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Invitrogen

          NKG2A/C/E Monoclonal Antibody (20d5), Functional Grade, eBioscience™

          3 Published Figures
          6 References
          View all (11) NKG2A/NKG2C/NKG2E antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Datasheet
          Protocols

          Cite NKG2A/C/E Monoclonal Antibody (20d5), Functional Grade, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (3)
          NKG2A/C/E Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          NKG2A/C/E Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NKG2A/C/E Antibody (16-5896-050) in Flow

          Staining of C57Bl/6 splenocytes with 0.5 µg of Rat IgG2a Isotype Control Purified (Product # 14-4321-82) (open histogram) or 0.125 µg of Anti-Mouse NKG2A/C/E Purified (filled histogram) followed by Anti-Rat IgG FITC (Product # 11-4811-85). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NKG2A/C/E Antibody in Flow Cytometry (Flow)
          NKG2A/C/E Antibody in Flow Cytometry (Flow)
          NKG2A/C/E Antibody in Flow Cytometry (Flow)
          NKG2A/C/E Antibody in Flow Cytometry (Flow)

          Product Details

          16-5896-050

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.06 µg/test
          View 4 publications 4 publications

          Functional Assay (Functional)

          Assay-Dependent
          -

          Control (Ctrl)

          Assay-Dependent
          -

          Miscellaneous PubMed (MISC)

          -
          View 1 publication 1 publication
          Product Specifications

          Published species

          Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), Functional Grade, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          20d5

          Conjugate

          Functional Grade Functional Grade Functional Grade
          • Biotin 
          • FITC
          • PerCP-eFluor 710
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Storage conditions

          4°C

          Shipping conditions

          Wet ice

          RRID

          AB_2866088

          Product Specific Information

          Description: The 20d5 monoclonal antibody reacts with the mouse NKG2A, C, and E. NKG2 molecules belong to a C-type lectin-like family of cell surface receptors expressed by mouse NK and NKT cell lineages. NKG2 molecules form heterodimeric complexes with CD94 and are responsible for recognition of non-classical MHC class I antigen Qa-1.

          When co-staining with NKG2A^B6 clone 16a11, it is important to stain for NKG2A-B6 first then subsequently with NKG2A/C/E, as steric hindrance has been observed if co-stained concurrently.

          Applications Reported: This 20d5 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This 20d5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.06 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Storage and handling: Use in a sterile environment.

          Filtration: 0.2 µm post-manufacturing filtered.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.

          Aggregation: Less than 10%, as determined by HPLC.

          Target Information

          Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity. The protein encoded by this gene belongs to the killer cell lectin-like receptor family, also called NKG2 family, which is a group of transmembrane proteins preferentially expressed in NK cells. This family of proteins is characterized by the type II membrane orientation and the presence of a C-type lectin domain. This protein forms a complex with another family member, KLRD1/CD94, and has been implicated in the recognition of the MHC class I HLA-E molecules in NK cells. The genes of NKG2 family members form a killer cell lectin-like receptor gene cluster on chromosome 12. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline