Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • KLRB1 (CD161) Antibodies

          Exalpha

          NKR-P1-A,B,C Monoclonal Antibody (1C10)

          View all (50) KLRB1 (CD161) antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite NKR-P1-A,B,C Monoclonal Antibody (1C10)

          Product Details

          X1166M

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          Flow Cytometry (Flow)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Mouse

          Host/Isotype

          Mouse / IgG

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1C10

          Immunogen

          Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized (BALB/c X C57BL/6)F1 (CbyB6F1) lymphokine-activated killer (LAK) cells and mouse myeloma cell line.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-2 mg/mL

          Purification

          Protein A/G

          Storage buffer

          PBS

          Contains

          0.08% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          This antibody binds to NKR-P1 receptors and induces release of cytokine in SJL/J NK cells.1.

          Target Information

          KLRB1 (Killer cell lectin like receptor subfamily B, member 1, CD-161) protein is classified as a type II membrane protein because it has an external C terminus. KLRB1 cell surface antigen is expressed by almost all NK cells and in a small subset of CD3+ve T cells. KLRB1 is a homodimeric cell surface protein, comprising two chains with molecular weights ranging from 40-44kDa. KLRB1 plays an inhibitory role on natural killer (NK) cells. Activation of KLRB1 leads to acid sphingomyelnase/SMPD1 stimulation and an increase in intracellular ceramide. Moreover, there is also an activation of AKT1/PKB and RPS6KA1/RSK1 kinase stimulation, and T cell proliferation by anti-CD3. KLRB1 acts as a lectin that binds to the terminal carbohydrate Gal-alpha(1,3)Gal epitope and to the N-acetyllactosamine epitope. KLRB1 also binds to CLEC2D/LLT1 as a ligand, and inhibits NK cell-mediated cytotoxicity as well as interferon-gamma secretion in target cells. The KLRB1 protein contains an extracellular domain with several motifs characteristic of C-type lectins, a transmembrane domain, and a cytoplasmic domain.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD161 antigen-like family member A; CD161 antigen-like family member B; CD161a; CD161b; immunoreceptor NKR-P1G; Inhibitory receptor NKR-P1B; killer cell lectin-like receptor subfamily A member 1 complex; Killer cell lectin-like receptor subfamily B member 1; killer cell lectin-like receptor subfamily B member 1 complex; Killer cell lectin-like receptor subfamily B member 1A; killer cell lectin-like receptor subfamily B member 1B allele A; Killer cell lectin-like receptor subfamily B member 1B allele B; killer cell lectin-like receptor subfamily B member 1D; Killer cell lectin-like receptor subfamily B member 1G; lectin-like transmembrane protein Nkrp1g; Ly-55A; ly-55b; Ly-55d; lymphocyte antigen 55; lymphocyte antigen 55 complex, locus A; lymphocyte antigen 55 complex, locus B; lymphocyte antigen 55 complex, locus D; Lymphocyte antigen 55A; lymphocyte antigen 55b; Lymphocyte antigen 55d; MGC138614; natural killer cell receptor; natural killer cell receptor protein NKR-P1A; natural killer cell receptor protein NKR-P1B; natural killer cell receptor-P1; Natural killer cell surface protein NKR-P1B allele B6; natural killer cell surface protein NKR-P1B allele SJL/BALB; Natural killer cell surface protein NKR-P1G; Natural killer cell surface protein P1-2; Natural killer lectin-like receptor 1E; natural killer lectin-like receptor protein 1 e; NKR-P1 2; NKR-P1 34; NKR-P1.7; NKR-P1A; NKR-P1D

          View more View less

          Gene Aliases: 4930431A04Rik; Gm4696; Klrb1; Klrb1a; Klrb1b; Klrb1d; Klrb1g; Klrb6; Ly-55; ly-55A; Ly55; Ly55a; Ly55b; Ly55d; NKR-P1.7; NKR-P1A; NKR-P1B; NKR-P1D; NKR-P1G; Nkrp-1e; Nkrp1-a; Nkrp1-b; NKRP12; Nkrp1a; Nkrp1b; Nkrp1d; Nkrp1g

          View more View less

          UniProt ID: (Mouse) Q0ZUP1, (Mouse) P27811, (Mouse) Q99JB4

          View more View less

          Entrez Gene ID: (Mouse) 100043861, (Mouse) 17057, (Mouse) 80782

          View more View less

          Function(s)
          receptor activity protein binding carbohydrate binding protein homodimerization activity
          Process(es)
          biological_process negative regulation of natural killer cell mediated cytotoxicity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0