Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Nkx2.5 Antibodies

          Abnova

          NKX2-5 Monoclonal Antibody (3E3)

          View all (23) Nkx2.5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite NKX2-5 Monoclonal Antibody (3E3)

          • Antibody Testing Data (1)
          NKX2-5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NKX2-5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NKX2-5 Antibody (H00001482-M08) in WB

          Western Blot analysis of NKX2-5 expression in transfected 293T cell line by NKX2-5 monoclonal antibody (M08), clone 3E3.

          Lane 1: NKX2-5 transfected lysate(34.92 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NKX2-5 Antibody in Western Blot (WB)

          Product Details

          H00001482-M08

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          ELISA (ELISA)

          1-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3E3

          Immunogen

          NKX2-5 (NP_004378, 1 a.a. approximately 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MFPSPALTPT PFSVKDILNL EQQQRSLAAA GELSARLEAT LAPSSCMLAA FKPEAYAGPE AAAPGLPELR AELGRAPSPA KCASAFPAAP AFYPRAYSDP DPAKDPRAEK KELCALQKAV ELEKTEADNA *

          Target Information

          This gene encodes a homeobox-containing transcription factor. This transcription factor functions in heart formation and development. Mutations in this gene cause atrial septal defect with atrioventricular conduction defect, and also tetralogy of Fallot, which are both heart malformation diseases. Mutations in this gene can also cause congenital hypothyroidism non-goitrous type 5, a non-autoimmune condition. Alternative splicing results in multiple transcript variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cardiac-specific homeobox; cardiac-specific homeobox 1; FLJ52202; FLJ97166; FLJ97195; FLJ97197; FLJ99536; Homeobox protein CSX; Homeobox protein NK-2 homolog E; homeobox protein NKX 2-5; Homeobox protein Nkx-2.5; NK2 transcription factor related locus 5; NK2 transcription factor related, locus 5; NKX 2-5; Nkx-25; tinman paralog

          View more View less

          Gene Aliases: CHNG5; CSX; CSX1; HLHS2; NKX2-5; NKX2.5; NKX2E; NKX4-1; VSD3

          View more View less

          UniProt ID: (Human) P52952

          View more View less

          Entrez Gene ID: (Human) 1482

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding RNA polymerase II transcription cofactor activity transcriptional activator activity, RNA polymerase II transcription factor binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding protein binding transcription factor binding serum response element binding protein homodimerization activity sequence-specific DNA binding transcription regulatory region DNA binding protein heterodimerization activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter vasculogenesis heart looping heart morphogenesis outflow tract septum morphogenesis cardiac conduction system development bundle of His development Purkinje myocyte differentiation cardiac ventricle formation right ventricular cardiac muscle tissue morphogenesis ventricular trabecula myocardium morphogenesis apoptotic process involved in heart morphogenesis septum secundum development proepicardium development pulmonary myocardium development transcription from RNA polymerase II promoter adult heart development positive regulation of cell proliferation negative regulation of cardiac muscle cell apoptotic process positive regulation of transcription via serum response element binding positive regulation of sodium ion transport negative regulation of myotube differentiation hemopoiesis cell differentiation BMP signaling pathway thyroid gland development embryonic heart tube development negative regulation of apoptotic process sarcomere organization positive regulation of neuron differentiation positive regulation of heart contraction negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter spleen development positive regulation of cardioblast differentiation ventricular cardiac myofibril assembly cardiac muscle cell differentiation cardiac muscle tissue morphogenesis atrial cardiac muscle cell development ventricular cardiac muscle cell development regulation of cardiac muscle contraction pharyngeal system development cardiac muscle cell proliferation regulation of cardiac muscle cell proliferation cardiac muscle contraction canonical Wnt signaling pathway positive regulation of transcription initiation from RNA polymerase II promoter heart trabecula formation ventricular septum morphogenesis atrial septum morphogenesis atrioventricular node cell development atrioventricular node cell fate commitment embryonic heart tube left/right pattern formation negative regulation of canonical Wnt signaling pathway regulation of cardiac conduction
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0