Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
This monoclonal antibody CSTEM30 reacts with human Neuroligin-4, X linked, also known as NLGN4X, NLGN4, Neuroligin-4, and Neuroligin 4X.
Applications Reported: This CSTEM30 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This CSTEM30 antibody has been pre-diluted and tested by flow cytometric analysis of human iPSCs. This may be used at 5 µL (0.06 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
Neuroligin-4, X linked (also known as NLGN4X, NLGN4, Neuroligin-4, and Neuroligin 4X) is a postsynaptic cell-adhesion molecule that interacts with presynaptic cell-surface neurexins. These interactions bridge together the synaptic cleft and have an essential role in synaptic transmissions between neurons. There are 5 neuroligin genes in humans: NLGN1, NLGN2, NLGN3, NLGN4X, and NLGN4Y. Mutations in NLGN3 and NLGN4X are linked to Autism Spectrum Disorders (ASD). The expression of neuroligin-4, X-linked has been reported on human pluripotent stem cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: HNLX; MGC22376; Neuroligin X; Neuroligin-4, X-linked; NLGN
Gene Aliases: ASPGX2; AUTSX2; HLNX; HNL4X; KIAA1260; NLGN4; NLGN4X; UNQ365/PRO701
UniProt ID: (Human) Q8N0W4
Entrez Gene ID: (Human) 57502
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support