Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive test controls include: U-87MG, U-251MG, HeLa, A-549, Mouse brain, Mouse testis, Rat brain, Rat testis. The target is usually found in the following locations: Cell junction, Cell membrane, Single-pass type I membrane protein, postsynaptic cell membrane, postsynaptic density, synapse.
Immunogen sequence: DMLPIWFTTS LDTLMTYVQD QNEDCLYLNI YVPMEDGTNI KRNADDITSN DHGEDKDIHE QNSKKPVMVY IHGGSYMEGT GNMIDGSILA SYGNVIVITI NYRLGILGMQ EARLCGSSKM FNYFKSPFTN LINFF
Neuroligins, such as NLGN4Y, are cell adhesion molecules present at the postsynaptic side of the synapse and may be essential for the formation of functional synapses (Jamain et al., 2003 [PubMed 12669065]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuroligin Y; Neuroligin-4, Y-linked
Gene Aliases: KIAA0951; NLGN4Y
UniProt ID: (Human) Q8NFZ3
Entrez Gene ID: (Human) 22829
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support