Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene encodes a member of the metallopeptidase M3 protein family that cleaves neurotensin at the Pro10-Tyr11 bond, leading to the formation of neurotensin(1-10) and neurotensin(11-13). The encoded protein is likely involved in the termination of the neurotensinergic signal in the central nervous system and in the gastrointestinal tract.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: angiotensin binding protein; Angiotensin-binding protein; endopeptidase 24.16; MEP; Microsomal endopeptidase; Mitochondrial oligopeptidase M; neurolysin (metallopeptidase M3 family); Neurolysin, mitochondrial; Neurotensin endopeptidase
Gene Aliases: 4930472G13Rik; AGTBP; C79345; EP24.16; KIAA1226; MEP; MOP; NLN
UniProt ID: (Human) Q9BYT8, (Mouse) Q91YP2, (Rat) P42676
Entrez Gene ID: (Human) 57486, (Mouse) 75805, (Rat) 117041
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support