Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: RERPFYGRLV KYMASGPVVA MVWQGLDVVR TSRALIGATN PADAPPGTIR GDFCIEVGKN LIHGSDSVES ARREIALWFR ADELLCWEDS AGHWLYE
NME3 mRNA is preferentially expressed at early stages of myeloid differentiation of highly purified CD34(+) cells. Its constitutive expression in a myeloid precursor line, which is growth-factor dependent for both proliferation and differentiation, results in inhibition of granulocytic differentiation induced by granulocyte colony-stimulating factor and causes apoptotic cell death. These results appear consistent with a role for the NME3 gene in normal hematopoiesis and raise the possibility that its overexpression contributes to differentiation arrest, a feature of blastic transformation in chronic myelogenous leukemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DR-nm23; NDK 3; NDP kinase 3; NDP kinase C; NDPKC; nm23-H3; non-metastatic cells 3, protein expressed in; Nucleoside diphosphate kinase 3; Nucleoside diphosphate kinase C
Gene Aliases: c371H6.2; DR-nm23; NDPK-C; NDPKC; NM23-H3; NM23H3; NME3
UniProt ID: (Human) Q13232
Entrez Gene ID: (Human) 4832
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support