Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MADESETAV KPPAPPLPQM MEGNGNGHEH CSDCENEEDN SYNRGGLSPA NDTGAKKKKK KQKKKKEKGS ETDSAQDQPV KMNSLPAERI QEIQKAIELF SVGQGPAKTM EEASKRSYQF WDTQPVPKLG EVVNTHGPVE PDKDNIRQEP YTLPQGFTWD ALDLGDRGVL KELYTLLNEN YVEDDDNMFR FDYSPEFLLW ALRPPGWLPQ WHCGVRVVSS RKLVGFISAI PANIHIYDTE KKMVEINFLC VHKKLRSKRV APVLIREITR RVHLEGIFQA VYTAGVVLPK PVGTCRYWHR SL (1-301 aa encoded by BC006569)
NMT1 is involved with Myristate, a rare 14-carbon saturated fatty acid, is cotranslationally attached by an amide linkage to the N-terminal glycine residue of cellular and viral proteins with diverse functions. N-myristoyltransferase (NMT EC 2.3.1.97) catalyzes the transfer of myristate from CoA to proteins. N-myristoylation appears to be irreversible and is required for full expression of the biologic activities of several N-myristoylated proteins, including the alpha subunit of the signal-transducing guanine nucleotide-binding protein (G protein) GO (GNAO1 MIM 139311).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alternative, short form NMT-S; Glycylpeptide N-tetradecanoyltransferase 1; HsNMT1; long form, NMT-L; Myristoyl-CoA:protein N-myristoyltransferase 1; NMT 1; Peptide N-myristoyltransferase 1; Protein-lysine myristoyltransferase NMT1; type I N-myristoyltransferase
Gene Aliases: AW536594; NMT; NMT1
UniProt ID: (Human) P30419, (Mouse) O70310, (Rat) Q8K1Q0
Entrez Gene ID: (Human) 4836, (Mouse) 18107, (Rat) 259274
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support