Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: IYNLLSRRFQ AAFQNVISSF HKQWHSQHDP QLPPAQRNIF LTECHFVELT EDIGPQFPCQ SSMHNSHLPA ALSSEQMSRT NYQSFHFNKT; Positive Samples: SGC-7901, Mouse uterus, Mouse small intestine, Rat uterus; Cellular Location: Cell membrane, Multi-pass membrane protein
This gene encodes a protein from the G-protein coupled receptor 1 family. This protein is a receptor for neuromedin U, which is a neuropeptide that is widely distributed in the gut and central nervous system. This receptor plays an important role in the regulation of food intake and body weight.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-protein coupled receptor FM-4; G-protein coupled receptor TGR-1; G-protein-coupled receptor FM-4; growth hormone secretagogue receptor family, member 4; Neuromedin-U receptor 2; NMU-R2
Gene Aliases: FM-4; FM4; NMU-R2; NMU2R; NMUR2; TGR-1; TGR1
UniProt ID: (Human) Q9GZQ4, (Rat) Q9ESQ4, (Mouse) Q8BZ39
Entrez Gene ID: (Human) 56923, (Rat) 64042, (Mouse) 216749
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support