Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
NOL10 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gm67; H_NH0074G24.1; Nucleolar protein 10; polyglutamine binding protein 5
Gene Aliases: NOL10; PQBP5
UniProt ID: (Human) Q9BSC4
Entrez Gene ID: (Human) 79954
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support