Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • NOL6 Antibodies

          Abnova

          NOL6 Polyclonal Antibody, MaxPab™

          View all (8) NOL6 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite NOL6 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          NOL6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NOL6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NOL6 Antibody (H00065083-B02P) in WB

          Western Blot analysis of NOL6 expression in transfected 293T cell line by NOL6 MaxPab polyclonal antibody. Lane 1: NOL6 transfected lysate(125.73 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NOL6 Antibody in Western Blot (WB)

          Product Details

          H00065083-B02P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NOL6 (AAH30139.1, 1 a.a. approximately 1143 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MGPAPAGEQL RGATGEPEVM EPALEGTGKE GKKASSRKRT LAEPPAKGLL QPVKLSRAEL YKEPTNEELN RLRETEILFH SSLLRLQVEE LLKEVRLSEK KKDRIDAFLR EVNQRVVRVP SVPETELTDQ AWLPAGVRVP LHQVPYAVKG CFRFLPPAQV TVVGSYLLGT CIRPDINVDV ALTMPREILQ DKDGLNQRYF RKRALYLAHL AHHLAQDPLF GSVCFSYTNG CHLKPSLLLR PRGKDERLVT VRLHPCPPPD FFRPCRLLPT KNNVRSAWYR GQSPAGDGSP EPPTPRYNTW VLQDTVLESH LQLLSTILSS AQGLKDGVAL LKVWLRQREL DKGQGGFTGF LVSMLVVFLV STRKIHTTMS GYQVLRSVLQ FLDLTVNGIS LCLSSDPSLP ALADFHQAFS VVFLDSSGHL NLCADVTAST YHQVQHEARL SMMLLDSRAD DGFHLLLMTP KPMIRAFDHV LHLRPLSRLQ AACHRLKLWP ELQDNGGDYV SAALGPLTTL LEQGLGARLN LLAHSRPPVP EWDISQDPPK HKDSGTLTLG LLLRPEGLTS VLELGPEADQ PEAAKFRQFW GSRSELRRFQ DGAIREAVVW EAASMSQKRL IPHQVVTHLL ALHADIPETC VHYVGGPLDA LIQGLKETSS TGEEALVAAV RCYDDLSRLL WGLEGLPLTV SAVQGAHPVL RYTEVFPPTP VRPAFSFYET LRERSSLLPR LDKPCPAYVE PMTVVCHLEG SGQWPQDAEA VQRVRAAFQL RLAELLTQQH GLQCRATATH TDVLKDGFVF RIRVAYQREP QILKEVQSPE GMISLRDTAA SLRLERDTRQ LPLLTSALHG LQQQHPAFSG VARLAKRWVR AQLLGEGFAD ESLDLVAAAL FLHPEPFTPP SSPQVGFLRF LFLVSTFDWK NNPLFVNLNN ELTVEEQVEI RSGFLAARAQ LPVMVIVTPQ DRKNSVWTQD GPSAQILQQL VVLAAEALPM LEKQLMDPRG PGDIRTVFRP PLDIYDVLIR LSPRHIPRHR QAVDSPAASF CRGLLSQPGP SSLMPVLGYD PPQLYLTQLR EAFGDLALFF YDQHGGEVIG VLWKPTSFQP QPFKASSTKG RMVMSRGGEL VMVPNVEAIL EDFAVLGEGL VQTVEARSER WTV

          Target Information

          The nucleolus is a dense subnuclear membraneless organelle that assembles around clusters of rRNA genes and functions in ribosome biogenesis. This gene encodes a nucleolar RNA-associated protein that is highly conserved between species. RNase treatment of permeabilized cells indicates that the nucleolar localization is RNA dependent. Further studies suggest that the protein is associated with ribosome biogenesis through an interaction with pre-rRNA primary transcripts. Alternative splicing has been observed at this locus and two splice variants encoding distinct isoforms have been identified.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Nrap; Nucleolar protein 6; nucleolar protein 6 (RNA-associated); nucleolar protein family 6 (RNA-associated); Nucleolar RNA-associated protein

          View more View less

          Gene Aliases: bA311H10.1; NOL6; NRAP; UTP22

          View more View less

          UniProt ID: (Human) Q9H6R4

          View more View less

          Entrez Gene ID: (Human) 65083

          View more View less

          Function(s)
          poly(A) RNA binding
          Process(es)
          rRNA processing tRNA export from nucleus
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0