Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NOL8 binds Ras-related GTP-binding proteins and plays a role in cell growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NOL8; Nucleolar protein 8; Nucleolar protein Nop132
Gene Aliases: 4921532D18Rik; 5730412B09Rik; bA62C3.3; bA62C3.4; C9orf34; NOL8; NOP132
UniProt ID: (Human) Q76FK4, (Mouse) Q3UHX0
Entrez Gene ID: (Human) 55035, (Rat) 361221, (Mouse) 70930
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support