Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Immunogen sequence: KSVQELRREQ QLAEIEARRQ EREKNGNEEG EERMTKPPVQ EMVDELQGPF SYDFSYWARS GEKITVTPSS KELLFYPPSM EAVVSGESCP GKLIEIHGKA GLFLEGQIHP ELEGVEIVIS EKGASSPLIT VFTDDKGAYS VGPLHSDLEY TVTSQKEGYV LTAVEGTIGD FKAYALAGVS FEIKAEDDQP LPGVLLSLSG GLFRSNLLTQ DNGILTFSNL SPGQYYFKPM MKEFRFEPSS QMIEVQEGQN LKITITGYRT AYSCYGTVSS LNGEPEQGVA MEAVGQNDCS IYGEDTVTDE E; Positive Samples: HT-29, A-431, MCF-7, Mouse brain, Mouse kidney, Mouse liver, Rat kidney; Cellular Location: Membrane, Single-pass type I membrane protein
This gene encodes a protein originally thought to be related to the collagenase gene family. This gene is one of three highly similar genes in a region of duplication located on the p arm of chromosome 16. These three genes encode closely related proteins that may have the same function. The protein encoded by one of these genes has been identified as part of a protein complex that participates in the Nodal signaling pathway during vertebrate development. Mutations in ABCC6, which is located nearby, rather than mutations in this gene are associated with pseudoxanthoma elasticum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BOS complex subunit NOMO1; Nodal modulator 1; nodal modulator 3; nomo1; pM5 protein; pM5 protein 3; pM5 protein, telomeric copy
Gene Aliases: D7Ertd156e; Nomo; NOMO1; PM5; RGD1305240
UniProt ID: (Human) Q15155, (Mouse) Q6GQT9
Entrez Gene ID: (Human) 23420, (Mouse) 211548, (Rat) 361578
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support