Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: QPLDLEPALP PGSRVILPTP EEQPLSRAAG RLSIHSLEAQ SLRCLQPFCT QDTRDRPFQA QAQESLDVLL RHPSGWWLVE NEDRQTAWFP APYLEEAAPG QGREGGPSLG SSGPQFCASR AYESSRADEL SVPAGARVRV LETSDRGWWL CRYGDRAGLL PAVLLRPEGL GALLSGTGFR GGDDPAGEAR GFPEPSQATA PPPTVPTRPS PGAIQSRCCT VTRRALERRP RRQGRPRGCV DSVPHPTTEQ
NADPH oxidases catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species to NOX and also target NOX to different subcellular compartments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC20258; NADPH oxidase organizer 1; Nox organizer 1; Sorting nexin-28
Gene Aliases: 2310034C04Rik; hslt; Noxo1; P41NOX; P41NOXA; P41NOXB; P41NOXC; Snx28
UniProt ID: (Mouse) Q8VCM2
Entrez Gene ID: (Mouse) 71893
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support