Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HSLEAQSLRC LQPFCTQDTR DRPFQAQAQE SLDVLLRHPS GWWLVENEDR
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 77%; Horse: 77%; Human: 100%
NADPH oxidases catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species to NOX and also target NOX to different subcellular compartments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC20258; NADPH oxidase organizer 1; NADPH oxidase regulatory protein; Nox organizer 1; Nox-organizing protein 1; regulatory protein P41NOX; SH3 and PX domain-containing protein 5
Gene Aliases: NOXO1; P41NOX; P41NOXA; P41NOXB; P41NOXC; SH3PXD5; SNX28
UniProt ID: (Human) Q8NFA2
Entrez Gene ID: (Human) 124056
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support