Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SLC17A1 Antibodies

          Proteintech

          NPT1 Polyclonal Antibody

          3 References
          View all (5) SLC17A1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite NPT1 Polyclonal Antibody

          • Antibody Testing Data (2)
          NPT1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NPT1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NPT1 Antibody (20751-1-AP) in WB

          Human placenta tissue were subjected to SDS PAGE followed by western blot with 20751-1-AP (NPT1 antibody) at dilution of 1:300 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NPT1 Antibody in Western Blot (WB)
          NPT1 Antibody in Western Blot (WB)

          Product Details

          20751-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2400
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NPT1 Fusion Protein Ag12819 (1-107 aa encoded by BC101745)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.3 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Western blot analysis using this antibody detected a major band around 45-55 kDa in kidney tissue.

          Immunogen sequence: MQMDNRLPP KKVPGFCSFR YGLSFLVHCC NVIITAQRAC LNLTMVVMVN STDPHGLPNT STKKLLDNIK NPMYNWSPDI QGIILSSTSY GVIIIQVPVG YFSGIYST (1-107 aa encoded by BC101745)

          Target Information

          SLC17A1 (Solute Carrier Family 17 Member 1) is a Protein Coding gene. Diseases associated with SLC17A1 include Gout and Hyperuricemia. Among its related pathways are Transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds and Uricosurics Pathway, Pharmacodynamics. Gene Ontology (GO) annotations related to this gene include symporter activity and phosphate ion transmembrane transporter activity. An important paralog of this gene is SLC17A2. Also, it is involved in cotransport of sodium and phosphate.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Na(+)/PI cotransporter 1; Na/Pi-4; Renal Na(+)-dependent phosphate cotransporter 1; Renal sodium-dependent phosphate transport protein 1; Renal sodium-phosphate transport protein 1; sodium phosphate transporter; Sodium-dependent phosphate transport protein 1; sodium-dependent phosphate/anion cotransporter; Sodium/phosphate cotransporter 1; sodium/phosphate type I cotransporter; solute carrier family 17 (organic anion transporter), member 1; solute carrier family 17 (sodium phosphate), member 1; solute carrier family 17 (sodium/hydrogen exchanger), member 1; solute carrier family 17 (vesicular glutamate transporter), member 1; Solute carrier family 17 member 1; solute carrier family 17 vesicular glutamate transporter), member 1

          View more View less

          Gene Aliases: NAPI-1; Napi1; NPT-1; NPT1; SLC17A1

          View more View less

          UniProt ID: (Human) Q14916, (Mouse) Q61983

          View more View less

          Entrez Gene ID: (Human) 6568, (Rat) 171080, (Mouse) 20504

          View more View less

          Function(s)
          sodium:phosphate symporter activity sodium-dependent phosphate transmembrane transporter activity phosphate ion transmembrane transporter activity symporter activity
          Process(es)
          phosphate ion transmembrane transport sodium ion transmembrane transport urate metabolic process transmembrane transport transport ion transport sodium ion transport sodium-dependent phosphate transport phosphate ion transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0