Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TLX Antibodies

          Bioss

          NR2E1/Tailless Polyclonal Antibody

          View all (17) TLX antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite NR2E1/Tailless Polyclonal Antibody

          Product Details

          BS-11202R

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:400
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human NR2E1/Tailless, amino acids 351-385.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          Nuclear receptor TLX, a NR2 hepatocyte NF4-like receptor, has been shown to affect anterior brain differentiation, retinal development, and vision. It has been suggested that TLX targets genes for RAR beta2 and Pax2 during retinal development. Homozygous TLX mutant mice are viable at birth but exhibit reduced forebrain development and are more aggressive than wild-type mice. Mice with deleted TLX locus exhibit cerebral hypoplasia, blindness, and extreme aggression (fierce (frc) phenotype). TLX expression has been documented in mouse brain and eye. ESTs have been isolated from normal human brain libraries.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: HTll; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; Nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; tailless homolog; Tll

          View more View less

          Gene Aliases: fierce; frc; Mtl1; Mtll; NR2E1; tailless; TLL; TLX; XTLL

          View more View less

          UniProt ID: (Human) Q9Y466, (Mouse) Q64104

          View more View less

          Entrez Gene ID: (Human) 7101, (Rat) 684085, (Mouse) 21907

          View more View less

          Function(s)
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding steroid hormone receptor activity zinc ion binding histone deacetylase binding sequence-specific DNA binding DNA binding enzyme binding transcription factor activity, sequence-specific DNA binding protein binding metal ion binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter behavioral fear response aggressive behavior transcription initiation from RNA polymerase II promoter nervous system development visual perception dentate gyrus development amygdala development olfactory bulb development layer formation in cerebral cortex forebrain generation of neurons cerebral cortex neuron differentiation anterior commissure morphogenesis extracellular matrix organization somatic stem cell population maintenance social behavior negative regulation of apoptotic process steroid hormone mediated signaling pathway cell fate commitment negative regulation of neuron differentiation positive regulation of angiogenesis positive regulation of cell cycle positive regulation of transcription from RNA polymerase II promoter negative regulation of astrocyte differentiation regulation of dendrite morphogenesis retina development in camera-type eye regulation of timing of neuron differentiation long-term synaptic potentiation regulation of cell migration involved in sprouting angiogenesis negative regulation of neural precursor cell proliferation positive regulation of neural precursor cell proliferation positive regulation of stem cell proliferation transcription, DNA-templated transcription from RNA polymerase II promoter brain development positive regulation of cell proliferation glial cell migration cerebral cortex development camera-type eye development regulation of cellular component organization regulation of transcription, DNA-templated multicellular organism development
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0