Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Sequence of this protein is as follows: MERDEPPPSG GGGGGGSAGF LEPPAALPPP PRNGFCQDEL AELDPGTNDR AEQRTCLICG DRATGLHYGI ISCEGCKGFF KRSICNKRVY RCSRDKNCVM SRKQRNRCQY CRLLKCLQMG MNRKAIREDG MPGGRNKSIG PVQISEEEIE RIMSGQEFEE EANHWSNHGD SDHSSPGNRA SESNQPSPGS TLSSRSVELN GFMAFREQYM GMSVPPHYQY IPHLFSYSGH SPLLPQQARS LDPQSYSLIH QLLSAEDLEP LGTPMLIEDG YAVTQAELFA LLCRLADELL FRQIAWIKKL PFFCELSIKD YTCLLSSTWQ ELILLSSLTV YSKQIFGELA DVTAKYSPSD EELHRFSDEG MEVIERLIYL YHKFHQLKVS NEEYACMKAI NFLNQDIRGL TSASQLEQLN KRYWYICQDF TEYKYTHQPN RFPDLMMCLP EIRYIAGKMV NVPLEQLPLL FKVVLHSCKT SVGKE
This gene encodes an orphan nuclear receptor which is a member of the nuclear hormone receptor family. Its expression pattern suggests that it may be involved in neurogenesis and germ cell development. The protein can homodimerize and bind DNA, but in vivo targets have not been identified. The gene expresses at least alternatively spliced transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GCNF; Germ cell nuclear factor; germ cell nuclear factor variant 1; h hRTR; Nuclear receptor subfamily 6 group A member 1; nuclear receptor subfamily 6, group A, member 1; retinoic acid receptor-related testis-associated receptor; Retinoid receptor-related testis-specific receptor; RTR
Gene Aliases: CT150; GCNF; GCNF1; hGCNF; hRTR; NR61; NR6A1; RTR
UniProt ID: (Human) Q8NHQ0
Entrez Gene ID: (Human) 2649
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support