Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PTPPAGAMST KQEARRDEGE ARTRGQEAQL RDRAHLSQQR RLKQATQFLH K
Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.
NRIP2 down-regulates transcriptional activation by nuclear receptors such as NR1F2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP761G1913; Nuclear receptor-interacting protein 2
Gene Aliases: NRIP2
UniProt ID: (Human) Q9BQI9
Entrez Gene ID: (Human) 83714
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support