Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
NSun6 is an RNA methyltransferase that catalyzes the methylation at position cytosine C72 in some tRNAs. NSUN6 has been reported to localize to the cytoplasm and largely colocalize with Golgi apparatus and pericentriolar matrix, indicating that NSUN6 modifies tRNAs in a late step in their biogenesis. NSun6 may also play a major role in bone metastasis and could be a valuable therapeutic target for bone metastasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARL5B antisense RNA 1; NOL1/NOP2/Sun and PUA domain-containing protein 1; NOL1/NOP2/Sun domain family member 6; NOL1/NOP2/Sun domain family, member 6; NOP2/Sun domain family, member 6; nucleolar protein (NOL1/NOP2/sun) and PUA domains 1; tRNA (cytosine(72)-C(5))-methyltransferase NSUN6
Gene Aliases: 4933414E04Rik; ARL5B-AS1; NOPD1; NSUN6
UniProt ID: (Human) Q8TEA1
Entrez Gene ID: (Human) 221078
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support