Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: WKDIVAAIQ HNYKMSAFKE NCGIYFPEIK RDPGRYLHSC PESVKKWLRQ LKNAGKILLL ITSSHSDYCR LLCEYILGND FTDLFDIVIT NALKPGFFSH LPSQRPFRTL ENDEEQEALP SLDKPGWYSQ GNAVHLYELL KKMTGKPEPK VVYFGDSMHS DIFPARHYSN WETVLILEEL RGDEGTRSQR PEESEPLEKK GKYEGPKAKP LNTSSKKWGS FFIDSVLGLE NTEDSLVYTW SCKRISTYST IAIPSIEAIA ELPLDYKFTR FSSSNSKTAG YYPNPPLVLS SDETLISK (1-296 aa encoded by BC015138)
While the exact function of the protein encoded by this gene is not known, it belongs to the 5'(3')-deoxyribonucleotidase family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-nucleotidase domain-containing protein 1; 5'-nucleotidase, cytosolic II-like 1 protein
Gene Aliases: C6orf200; LP2642; NT5C2L1; NT5DC1
UniProt ID: (Human) Q5TFE4
Entrez Gene ID: (Human) 221294
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support