Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDVSLCPAKC SFWRIFLLGS VWLDYVGSVL ACPANCVCSK TEINCRRPDD GNLFPLLEGQ DSGNSNGNAS INITDISRNI TSIHIENWRS LHTLNAVDME LYTGLQKLTI KNSGLRSIQP RAFAKNPHLR YINLSSNRLT TLSWQLFQTL SLRELQLEQN FFNCSCDIRW MQLWQEQGEA KLNSQNLYCI NADGSQLPLF RMNISQCDLP EISVSHVNLT VREGDNAVIT CNGSGSPLPD VDWIVTGLQS INTHQTNLNW TNVHAINLTL VNVTSEDNGF TLTCIAENVV GMSNASVALT VYYPPRVVSL EEPELRLEHC IEFVVRGNPP PTLHWLHNGQ PLRESKIIHV EYYQEGEISE GCLLFNKPTH YNNGNYTLIA KNPLGTANQT INGHFLKEPF PESTDNFILF DEVSPTPPIT VTHKPEEDTF GVSIAVGLAA FACVLLVVLF VMINKYGRRS KFGMKGPVAV ISGEEDSASP LHHINHGITT PSSLDAGPDT VVIGMTRIPV IENPQYFRQG HNCHKPDTWV FSNIDNHGIL NLKDNRDHLV PSTHYIYEEP EVQSGEVSYP RSHGFREIML NPISLPGHSK PLNHGIYVED VNVYFSKGRH GF
NTRK3 (TRKC) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. Known substrates for the TRK receptors are SHC, PI-3 kinase, and PLCG1. The different isoforms do not have identical signaling properties. The protein is widely expressed, mainly in the nervous tissue. The isoform B is expressed in a relatively large amount in the adult brain comparatively to fetal brain. TRKC is subject to ligand-mediated auto-phosphorylation. The protein structure contains 2 immunoglobulin-like C2-type domains and 2 leucine-rich (LRR) repeats.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ETS related protein-neurotrophic receptor tyrosine kinase fusion protein; ETV6-NTRK3 fusion; GP145-TrkC; Neurotrophic tyrosine kinase receptor type 3; neurotrophic tyrosine kinase, receptor, type 3; NT-3 growth factor receptor; Trk-C; TrkC tyrosine kinase; tyrosine kinase receptor C
Gene Aliases: gp145(trkC); GP145-TrkC; NTRK3; TRKC
UniProt ID: (Human) Q16288
Entrez Gene ID: (Human) 4916
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support