Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEGAAAPVAG DRPDLGLGAP GSPREAVAGA TAALEPRKPH GVKRHHHKHN LKHRYELQET LGKGTYGKVK RATERFSGRV VAIKSIRKDK IKDEQDMVHI RREIEIMSSL NHPHIISIYE VFENKDKIVI IMEYASKGEL YDYISERRRL SERETRHFFR QIVSAVHYCH KNGVVHRDLK LENILLDDNC NIKIADFGLS NLYQKDKFLQ TFCGSPLYAS PEIVNGRPYR GPEVDSWALG VLLYTLVYGT MPFDGFDHKN LIRQISSGEY REPTQPSDAR GLIRWMLMVN PDRRATIEDI ANHWWVNWGY KSSVCDCDAL HDSESPLLAR IIDWHHRSTG LQADTEAKMK GLAKPTTSEV MLERQRSLKK SKKENDFAQS GQDAVPESPS KLSSKRPKGI LKKRSNSEHR SHSTGFIEGV VGPALPSTFK MEQDLCRTGV LLPSSPEAEV PGKLSPKQSA TMPKKGILKK TQQRESGYYS SPERSESSEL LDSNDVMGSS IPSPSPPDPA RVTSHSLSCR RKGILKHSSK YSAGTMDPAL VSPEMPTLES LSEPGVPAEG LSRSYSRPSS VISDDSVLSS DSFDLLDLQE NRPARQRIRS CVSAENFLQI QDFEGLQNRP RPQYLKRYRN RLADSSFSLL TDMDDVTQVY KQALEICSKL N
NUAK1 (ARK5) is a serine/threonine kinase that plays a role in many cellular functions. The overexpression of this kinase has been associated with tumor invasion and metastasis (colorectal neoplasm). Acts as a regulator of cellular senescence and cellular ploidy by mediating phosphorylation of 'Ser-464' of LATS1, thereby controlling its stability. Controls cell adhesion by regulating activity of the myosin protein phosphatase 1 (PP1) complex. Acts by mediating phosphorylation of PPP1R12A subunit of myosin PP1: phosphorylated PPP1R12A then interacts with 14-3-3, leading to reduced dephosphorylation of myosin MLC2 by myosin PP1. May be involved in DNA damage response: phosphorylates p53/TP53 at 'Ser-15' and 'Ser-392' and is recruited to the CDKN1A/WAF1 promoter to participate in transcription activation by p53/TP53. May also act as a tumor malignancy-associated factor by promoting tumor invasion and metastasis under regulation and phosphorylation by AKT1. Suppresses Fas-induced apoptosis by mediating phosphorylation of CASP6, thereby suppressing the activation of the caspase and the subsequent cleavage of CFLAR. Regulates UV radiation-induced DNA damage response mediated by CDKN1A. In association with STK11, phosphorylates CDKN1A in response to UV radiation and contributes to its degradation which is necessary for optimal DNA repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AMP-activated protein kinase family member 5; AMPK-related protein kinase 5; ARK5; NUAK family SNF1-like kinase 1; NUAK family, SNF1-like kinase, 1; Omphalocele kinase 1
Gene Aliases: ARK5; KIAA0537; NUAK1; OMPHK1
UniProt ID: (Human) O60285
Entrez Gene ID: (Human) 9891
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support