Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVCGRQLSG AGSETLKQRR TQIMSRGLPK QKPIEGVKQV IVVASGKGGV GKSTTAVNLA LALAANDSSK AIGLLDVDVY GPSVPKMMNL KGNPELSQSN LMRPLLNYGI ACMSMGFLVE ESEPVVWRGL MVMSAIEKLL RQVDWGQLDY LVVDMPPGTG DVQLSVSQTI PITGAVIVST PQDIALMDAH KGAEMFRRVH VPVLGLVQNM SVFQCPKCKH KTHIFGADGA RKLAQTLGLE VLGDIPLHLN IREASDTGQP IVFSQPESDE AKAYLRIAVE VVRRLPSPSE (1-289 aa encoded by BC024919)
Required for the assembly of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). May deliver of one or more Fe-S clusters to complex I subunits.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: huInd1; IND1 homolog; Iron-sulfur cluster transfer protein NUBPL; iron-sulfur protein NUBPL; iron-sulfur protein required for NADH dehydrogenase; nucleotide binding protein-like; Nucleotide-binding protein-like
Gene Aliases: 2410170E07Rik; C14orf127; huInd1; IND1; NUBPL; RGD1307232
UniProt ID: (Human) Q8TB37, (Mouse) Q9CWD8
Entrez Gene ID: (Human) 80224, (Mouse) 76826, (Rat) 299008
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support