Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAHGSQEAE APGAVAGAAE VPREPPILPR IQEQFQKNPD SYNGAVRENY TWSQDYTDLE VRVPVPKHVV KGKQVSVALS SSSIRVAMLE ENGERVLMEG KLTHKINTES SLWSLEPGKC VLVNLSKVGE YWWNAILEGE EPIDIDKINK ERSMATVDEE EQAVLDRLTF DYHQKLQGKP QSHELKVHEM LKKGWDAEGS PFRGQRFDPA MFNISPGAVQ F (1-220 aa encoded by BC003691)
NUDCD3 gene ontology annotations related to this gene include cilium assembly; developmental process; protein folding; protein localization to pericentriolar material.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NudC domain-containing protein 3; NudC-like protein
Gene Aliases: AI427847; BC024322; KIAA1068; mKIAA1068; NUDCD3; NudCL
UniProt ID: (Human) Q8IVD9, (Mouse) Q8R1N4
Entrez Gene ID: (Human) 23386, (Mouse) 209586
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support