Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: QEWEAHRNLS QLPNFAFSVP LAYFLLSQQT DLPECEQSSA RQKASLLIQQ ALTMFPGVLL PLLESCSVRP DASVSSHRFF GPNAEISQ
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
TCF25 is a member of the basic helix-loop-helix (bHLH) family of transcription factors that are important in embryonic development (Steen and Lindholm, 2008 [PubMed 18068114]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nuclear localized protein 1; NULP1; Ribosome quality control complex subunit TCF25; TCF-25; Transcription factor 25; transcription factor 25 (basic helix-loop-helix)
Gene Aliases: 1100001J13Rik; 1810041K11Rik; D8Ertd325e; FKSG26; hKIAA1049; Hulp1; KIAA1049; mKIAA1049; NULP1; PRO2620; RGD1309054; TCF25
UniProt ID: (Human) Q9BQ70, (Mouse) Q8R3L2
Entrez Gene ID: (Human) 22980, (Rat) 292082, (Mouse) 66855
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support