Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEELDGEPTV TLIPGVNSKK NQMYFDWGPG EMLVCETSFN KKEKSEMVPS CPFIYIIRKD VDVYSQILRK LFNESHGIFL GLQRIDEELT GKSRKSQLVR VSKNYRSVIR ACMEEMHQVA IAAKDPANGR QFSSQVSILS AMELIWNLCE ILFIEVAPAG PLLLHLLDWV RLHVCEVDSL SADVLGSENP SKHDSFWNLV TILVLQGRLD EARQMLSKEA DASPASAGIC RIMGDLMRTM PILSPGNTQT LTELELKWQH WHEECERYLQ DSTFATSPHL ESLLKIMLGD EAALLEQKEL LSNWYHFLVT RLLYSNPTVK PIDLHYYAQS SLDLFLGGES SPEPLDNILL AAFEFDIHQV IKECSIALSN WWFVAHLTDL LDHCKLLQSH NLYFGSNMRE FLLLEYASGL FAHPSLWQLG VDYFDYCPEL GRVSLELHIE RIPLNTEQKA LKVLRICEQR QMTEQVRSIC KILAMKAVRN NRLGSALSWS IRAKDAAFAT LVSDRFLRDY CERGCFSDLD LIDNLGPAMM LSDRLTFLGK YREFHRMYGE KRFADAASLL LSLMTSRIAP RSFWMTLLTD ALPLLEQKQV IFSAEQTYEL MRCLEDLTSR RPVHGESDTE QLQDDDIETT KVEMLRLSLA RNLARAIIRE GSLEGS
Bidirectional transport of macromolecules between the cytoplasm and nucleus occurs through nuclear pore complexes (NPCs) embedded in the nuclear envelope. NPCs are composed of subcomplexes, and NUP85 is part of one such subcomplex, Nup107-160.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 85 kDa nucleoporin; FLJ12549; FROUNT; Nuclear pore complex protein Nup85; nucleoporin 85kDa; Nucleoporin Nup75; Nucleoporin Nup85; PCN; PCNT; Pericentrin-1
Gene Aliases: FROUNT; NUP75; NUP85; PCNT1
UniProt ID: (Human) Q9BW27
Entrez Gene ID: (Human) 79902
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support