Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Netrin-G1 ligand (NGL1) is a transmembrane protein containing leucine-rich repeat (LRR) and immunoglobulin (Ig) domains that specifically interact with netrin-G1 through its LRR region. Whereas netrin-G1 is expressed highly in mouse thalamic axons, NGL1 is most abundant in the striatum and the cerebral cortex. NGL1 is implicated in facilitating axonal growth of the vertebrate CNS. Membrane-bound netrin-G1 acts as an axonal receptor, whereas the LRR-containing transmembrane protein NGL1 acts as a ligand in promoting the growth of thalamic axons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epididymis tissue protein Li 131P; HGNC:8029; netrin 1, mouse, homolog of; Netrin-1; Netrin1
Gene Aliases: AI561871; BC019633; Netrin-1; NTN1; NTN1L
UniProt ID: (Human) O95631, (Mouse) O09118
Entrez Gene ID: (Human) 9423, (Mouse) 18208, (Rat) 114523
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support