Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
NKX2-3 encodes a homeodomain-containing transcription factor. The encoded protein is a member of the NKX family of homeodomain transcription factors. Studies of similar proteins in mouse and rat have indicated a potential role in cellular differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein NK-2 homolog C; Homeobox protein Nkx-2.3; NK2 transcription factor related, locus 3
Gene Aliases: CSX3; NK2.3; NKX2-3; NKX2.3; NKX23; NKX2C; NKX4-3
UniProt ID: (Human) Q8TAU0
Entrez Gene ID: (Human) 159296
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support