Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 83%
This gene encodes a member of the NK family of homeobox-containing proteins. The encoded protein may play a role in skeletal development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bagpipe homeobox protein homolog 1; Homeobox protein NK-3 homolog B; Homeobox protein Nkx-3.2
Gene Aliases: BAPX1; NKX3-2; NKX3.2; NKX3B; SMMD
UniProt ID: (Human) P78367
Entrez Gene ID: (Human) 579
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support