Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: SSSPSAQVIG LKNALSSALA QNTDLKERLR RIHAESLLLD SPAVAKSGDN LAEENSRDEN RALVHQLSNE SRLSITDSLS EFFDAQEVLL SPSSSENEIS DDDSYVSDIS DNLSLDNLSN DLDNERQTLG PVLDSGREAK SRRRTCLPAP C
ZNF703 belongs to the Elbow/Noc family. It contains 1 C2H2-type zinc finger.ZNF703 may function as a transcriptional repressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ORP-3; OSBP-related protein 3; oxysterol binding protein-like 3; Oxysterol-binding protein-related protein 3
Gene Aliases: 1200014M06Rik; 6720421I08Rik; A530055M08; KIAA0704; ORP-3; ORP3; OSBP3; OSBPL3; RGD1564287
UniProt ID: (Human) Q9H4L5, (Mouse) Q9DBS9
Entrez Gene ID: (Human) 26031, (Rat) 362360, (Mouse) 71720
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support