Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: WFEKALRDK KGFIIKQMKE DGACLFRAVA DQVYGDQDMH EVVRKHCMDY LMKNADYFSN YVTEDFTTYI NRKRKNNCHG NHIEMQAMAE MYNRPVEVYQ YSTEPINTFH GIHQNEDEPI RVSYHRNIHY NSVVNPNKAT IGVGLGLPSF KPGFAEQSLM KNAIKTSEES WIEQQMLEDK KRATDWEATN EAIEEQVARE SYLQWLRDQE KQARQVRGPS QPRKASATCS SATAAASSGL EEWTSRSPRQ RSSASSPEHP ELHAELGMKP PSPGTVLALA KPPSPCAPGT SSQFSAGADR ATSPLVSLYP ALECRALIQQ MSPSAFGLND WDDDEILASV LAVSQQEYLD SMKKNKVPCR C (202-561 aa encoded by BC028225)
This gene encodes a class 1 aminoacyl-tRNA synthetase, cysteinyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. This gene is one of several located near the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: deubiquinating enzyme A; Deubiquitinating enzyme A; DUBA; OTU domain containing 5; OTU domain-containing protein 5
Gene Aliases: AA407879; AI553596; BB114028; DUBA; DXImx46e; OTUD5; Sfc7
UniProt ID: (Human) Q96G74, (Rat) Q2YDU3, (Mouse) Q3U2S4
Entrez Gene ID: (Human) 55593, (Rat) 363452, (Mouse) 54644
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support