Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MGHPPLEFS DCYLDSPDFR ERLKCYEQEL ERTNKFIKDV IKDGNALISA MRNYSSAVQK FSQTLQSFQF DFIGDTLTDD EINIAESFKE FAELLNEVEN ERMMMVHNAS DLLIKPLENF RKEQIGFTKE RKKKFEKDGE RFYSLLDRHL HLSSKKKESQ LQEADLQVDK ERHNFFESSL DYVYQIQEVQ ESKKFNIVEP VLAFLHSLFI SNSLTVELTQ DFLPYKQQLQ LSLQNTRNHF SSTREEMEEL KKRMKEAPQT CKLPGQPTIE GYLYTQEKCV WGHRGIHWVS ISQELLPLVG CEFWAPLLFI DP (1-311 aa encoded by BC059393)
Stimulates GTP hydrolysis of members of the Rho family. Its action on RHOA activity and signaling is implicated in growth and stabilization of dendritic spines, and therefore in synaptic function. Critical for the stabilization of AMPA receptors at postsynaptic sites. Critical for the regulation of synaptic vesicle endocytosis at presynaptic terminals. Required for the localization of NR1D1 to dendrites, can suppress its repressor activity and protect it from proteasomal degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mental retardation, X-linked 60; Oligophrenin-1; oligophrenin-1, Rho-GTPase activating protein
Gene Aliases: ARHGAP41; C130037N19Rik; MRX60; OPHN1; OPN1
UniProt ID: (Human) O60890, (Rat) P0CAX5, (Mouse) Q99J31
Entrez Gene ID: (Human) 4983, (Rat) 312108, (Mouse) 94190
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support