Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Osenses
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in rat and human.
Specificity of this antibody: Orai2.
Antigen stimulation of immune cells triggers Ca++ entry through Ca++ release-activated Ca++ (CRAC) channels. ORAI2 is one of two mammalian homologs to ORAI1, a recently identified four-transmembrane spanning protein that is an essential component of CRAC. Like ORAI1, ORAI2 has been shown to function as a highly selective Ca++ plasma membrane channel that is gated through interactions with STIM1, the store-activated endoplasmic reticulum Ca++ sensor, although at a lesser efficacy than ORAI1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium release-activated calcium channel 2; calcium release-activated calcium modulator 2 protein; CAP-binding protein complex interacting protein 2; CAP-binding protein complex-interacting protein 2; H_NH0514P08.8; Protein orai-2; putative protein ORAI2-2; Transmembrane protein 142B
Gene Aliases: A730041O15Rik; C7orf19; CBCIP2; MEM142B; ORAI2; PP1729; RGD1310213; TMEM142B
UniProt ID: (Human) Q96SN7, (Mouse) Q8BH10
Entrez Gene ID: (Human) 80228, (Mouse) 269717, (Rat) 304592
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support