Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • P2X7 Antibodies

          Alomone Labs, LTD

          P2X7 Receptor (extracellular) Polyclonal Antibody, Atto™ 633

          View all (28) P2X7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite P2X7 Receptor (extracellular) Polyclonal Antibody, Atto™ 633

          • Antibody Testing Data (2)
          P2X7 Receptor (extracellular) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          P2X7 Receptor (extracellular) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          P2X7 Receptor (extracellular) Antibody (APR-008-FR-50UL) in ICC/IF

          Expression of P2RX7 in rat brain glioma (C6) cells - Cell surface detection of P2RX7 in intact living rat C6 cells. A. Extracellular staining of cells with Anti-P2X7 Receptor (extracellular)-ATTO Fluor-633 Antibody (#APR-008-FR), (1:25, purple). B. Merged image of A and nuclear staining using DAPI as the counterstain (blue). C. Merged image of B with cells live image. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          P2X7 Receptor (extracellular) Antibody in Immunocytochemistry (ICC/IF)
          P2X7 Receptor (extracellular) Antibody in Flow Cytometry (Flow)

          Product Details

          APR-008-FR-50UL

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:25
          -

          Flow Cytometry (Flow)

          10 µg/1x10^6 cells
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KKGWMDPQSKGIQTGRC, corresponding to amino acid residues 136-152 of mouse P2X7 receptor, Extracellular loop
          View immunogen

          Conjugate

          Atto™ 633 Atto™ 633 Atto™ 633

          Excitation/Emission Max

          630/655 nm

          Form

          Lyophilized

          Concentration

          1 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitution: 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C, protected from the light, for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 X g 5 min).

          Target Information

          The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and is responsible for ATP-dependent lysis of macrophages through the formation of membrane pores permeable to large molecules. Activation of this nuclear receptor by ATP in the cytoplasm may be a mechanism by which cellular activity can be coupled to changes in gene expression. Multiple alternatively spliced variants have been identified, most of which fit nonsense-mediated decay criteria.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATP receptor; MGC20089; P2X purinoceptor 7; p2X receptor 7; P2X7; P2X7 purinoceptor; P2X7 receptor; P2Z; P2Z receptor; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 7; purinergic receptor P2X, ligand-gated ion channel, 7; purinergic receptor P2X7; purinergic receptor P2X7 variant A

          View more View less

          Gene Aliases: AI467586; P2RX7; P2X(7); P2X7; P2X7R

          View more View less

          UniProt ID: (Human) Q99572, (Rat) Q64663, (Mouse) Q9Z1M0

          View more View less

          Entrez Gene ID: (Human) 5027, (Rat) 29665, (Mouse) 18439

          View more View less

          Function(s)
          lipopolysaccharide binding purinergic nucleotide receptor activity extracellular ATP-gated cation channel activity receptor binding protein binding ATP binding protein homodimerization activity magnesium ion binding receptor activity copper ion binding zinc ion binding channel activity protein serine/threonine kinase activator activity scaffold protein binding ion channel activity
          Process(es)
          activation of MAPK activity cell morphogenesis phagolysosome assembly positive regulation of T cell mediated cytotoxicity positive regulation of protein phosphorylation regulation of sodium ion transport protein phosphorylation membrane protein ectodomain proteolysis phospholipid transfer to membrane cation transport calcium ion transport cell volume homeostasis membrane budding inflammatory response mitochondrion organization plasma membrane organization cell surface receptor signaling pathway response to mechanical stimulus response to bacterium response to organic substance response to zinc ion gene expression positive regulation of calcium ion transport into cytosol programmed cell death positive regulation of glutamate secretion positive regulation of gamma-aminobutyric acid secretion response to organic cyclic compound organic cation transport synaptic vesicle exocytosis protein processing neuronal action potential sensory perception of pain cytolysis positive regulation of bone mineralization cellular response to extracellular stimulus bleb assembly positive regulation of prostaglandin secretion response to lipopolysaccharide positive regulation of interleukin-1 beta production positive regulation of interleukin-6 production collagen metabolic process response to ATP response to fluid shear stress purinergic nucleotide receptor signaling pathway T cell proliferation response to drug T cell homeostasis positive regulation of apoptotic process positive regulation of catalytic activity NAD transport negative regulation of MAPK cascade positive regulation of MAPK cascade multicellular organismal protein catabolic process phospholipid translocation positive regulation of ossification negative regulation of bone resorption positive regulation of cytolysis ceramide biosynthetic process pore complex assembly skeletal system morphogenesis homeostasis of number of cells within a tissue positive regulation of protein secretion positive regulation of cytokine secretion positive regulation of interleukin-1 alpha secretion positive regulation of interleukin-1 beta secretion defense response to Gram-positive bacterium release of sequestered calcium ion into cytosol protein oligomerization positive regulation of cytoskeleton organization response to calcium ion response to electrical stimulus membrane depolarization positive regulation of mitochondrial depolarization positive regulation of lymphocyte apoptotic process cellular response to organic cyclic compound reactive oxygen species metabolic process apoptotic signaling pathway extrinsic apoptotic signaling pathway cation transmembrane transport transport ion transport positive regulation of gene expression phospholipid scrambling positive regulation of ion transmembrane transport negative regulation of cell volume positive regulation of glycolytic process cellular response to dsRNA positive regulation of bleb assembly blood coagulation regulation of killing of cells of other organism
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0