Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: YPTLSEYVQD FLNHLTEQPG SFETEIEQFA ETLNGCVTTD DALQELVELI YQQATSIPNF SYMGARLCNY LSHHLTISPQ SGNFRQLLLQ RCRTEYEVKD QAAKGDEVTR
The protein encoded by this gene interacts with poly(A)-binding protein and with the cap-binding complex eIF4A. It is involved in translational initiation and protein biosynthesis. Overexpression of this gene in COS7 cells stimulates translation. Alternative splicing occurs at this locus and three transcript variants encoding three distinct isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PABC1-interacting protein 1; PABP-interacting protein 1; PAIP-1; poly(A)-binding protein-interacting protein 1; polyadenylate binding protein-interacting protein 1; Polyadenylate-binding protein-interacting protein 1
Gene Aliases: PAIP1
UniProt ID: (Human) Q9H074
Entrez Gene ID: (Human) 10605
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support