Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MFRKKKKKRP EISAPQNFQH RVHTSFDPKE GKFVGLPPQW QNILDTLRRP KPVVDPSRIT RVQLQPMKTV VRGSAMPVDG YISGLLNDIQ KLSVISSNTL RGRSPTSRRR AQSLGLLGDE HWATDPDMYL QSPQSERTDP HGLYLSCNGG TPAGHKQMPW PEPQSPRVLP NGLAAKAQSL GPAEFQGASQ RCLQLGACLQ SSPPGASPPT GTNRHGMKAA KHGSEEARPQ SCLVGSATGR PGGEGSPSPK TRESSLKRRL FRSMFLSTAA; Positive Samples: Mouse brain, Mouse spinal cord, Mouse heart, Rat brain, Rat spinal cord; Cellular Location: Cytoplasm, Nucleus
PAK6 is a serine/threonine kinase expressed in the testis and prostate. It associates with both the estrogen and androgen receptors. The p21-activated kinases (PAKs) are serine-threonine kinases that bind to the active forms of Cdc42 and Rac. They are divided into two groups, the first of which include PAK1, 2 and 3, and can be activated by Cdc42/Rac binding. Group 1 PAKs contain an autoinhibitory domain whose activity is regulated by Cdc42/Rac binding. The group 1 PAKs are known to be involved in cellular processes such as gene transcription, apoptosis, and cell morphology and motility. Much less is known about the second group, which includes PAK4, 5 and 6. These proteins are not activated by Cdc42/Rac binding. Later experiments have shown it to be activated by MAP kinase kinase 6 and p38 MAP kinase, suggesting that PAK6 may play a role in the cellular response to stress-related signals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: p21 (CDKN1A)-activated kinase 6; p21 protein (Cdc42/Rac)-activated kinase 6; p21(CDKN1A)-activated kinase 6; p21-activated kinase 6; PAK-5; PAK-6; Serine/threonine-protein kinase PAK 6
Gene Aliases: 4732456M09; PAK5; PAK6
UniProt ID: (Human) Q9NQU5, (Mouse) Q3ULB5
Entrez Gene ID: (Human) 56924, (Mouse) 214230, (Rat) 296078
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support