Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total PALMD.
PALMD gene ontology annotations related to this gene include regulation of cell shape.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Palmdelphin; Paralemmin-like protein; paralemmin-related; paralemnin-like protein
Gene Aliases: 4631423C22Rik; C1orf11; PALMD; PALML
UniProt ID: (Human) Q9NP74, (Mouse) Q9JHU2, (Rat) Q4KM62
Entrez Gene ID: (Human) 54873, (Mouse) 114301, (Rat) 310811
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support