Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKLINGKKQT FPWFGMDIGG TLVKLVYFEP KDITAEEEQE EVENLKSIRK YLTSNTAYGK TGIRDVHLEL KNLTMCGRKG NLHFIRFPSC AMHRFIQMGS EKNFSSLHTT LCATGGGAFK FEEDFRMIAD LQLHKLDELD CLIQGLLYVD SVGFNGKPEC YYFENPTNPE LCQKKPYCLD NPYPMLLVNM GSGVSILAVY SKDNYKRVTG TSLGGGTFLG LCCLLTGCET FEEALEMAAK GDSTNVDKLV KDIYGGDYER FGLQGSAVAS SFGNMMSKEK RDSISKEDLA RATLVTITNN IGSIARMCAL NENIDRVVFV GNFLRINMVS MKLLAYAMDF WSKGQLKALF LEHEGYFGAV GALLELFKMT DDK
PANK1 belongs to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPanK; hPanK1; Pantothenate kinase 1; Pantothenic acid kinase 1
Gene Aliases: PANK; PANK1
UniProt ID: (Human) Q8TE04
Entrez Gene ID: (Human) 53354
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support