Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Sequence of this protein is as follows: MSGIKKQKTE NQQKSTNVVY QAHHVSRNKR GQVVGTRGGF RGCTVWLTGL SGAGKTTISF ALEEYLVSHA IPCYSLDGDN VRHGLNRNLG FSPGDREENI RRIAEVAKLF ADAGLVCITS FISPFAKDRE NARKIHESAG LPFFEIFVDA PLNICESRDV KGLYKRARAG EIKGFTGIDS DYEKPETPER VLKTNLSTVS DCVHQVVELL QEQNIVPYTI IKDIHELFVP ENKLDHVRAE AETLPSLSIT KLDLQWVQVL SEGWATPLKG FMREKEYLQV MHFDTLLDDG VINMSIPIVL PVSAEDKTRL EGCSKFVLAH GGRRVAILRD AEFYEHRKEE RCSRVWGTTC TKHPHIKMVM ESGDWLVGGD LQVLEKIRWN DGLDQYRLTP LELKQKCKEM NADAVFAFQL RNPVHNGHAL LMQDTRRRLL ERGYKHPVLL LHPLGGWTKD DDVPLDWRMK QHAAVLEEGV LDPKSTIVAI FPSPMLYAGP TEVQWHCRSR MIAGANFYIV GRDPAGMPHP ETKKDLYEPT HGGKVLSMAP GLTSVEIIPF RVAAYNKAKK AMDFYDPARH NEFDFISGTR MRKLAREGEN PPDGFMAPKA WKVLTDYYRS LEKN
Sulfation is a common modification of endogenous (lipids, proteins, and carbohydrates) and exogenous (xenobiotics and drugs) compounds. In mammals, the sulfate source is 3'-phosphoadenosine 5'-phosphosulfate (PAPS), created from ATP and inorganic sulfate. Two different tissue isoforms encoded by different genes synthesize PAPS. This gene encodes one of the two PAPS synthetases. Defects in this gene cause the Pakistani type of spondyloepimetaphyseal dysplasia. Two alternatively spliced transcript variants that encode different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-prime-phosphoadenosine 5-prime-phosphosulfate synthase 2; adenosine 5'-phosphosulfate kinase; adenylyl-sulfate kinase; ATP sulfurylase/adenosine 5'-phosphosulfate kinase; ATP sulfurylase/APS kinase 2; Bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2; bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthethase 2; PAPS synthase 2; PAPS synthetase 2; PAPS synthethase 2; SK 2; sulfate adenylyltransferase; Sulfurylase kinase 2
Gene Aliases: ATPSK2; BCYM4; PAPSS2; SK2
UniProt ID: (Human) O95340
Entrez Gene ID: (Human) 9060
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support