Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MAWRGWAQRG WGCGQAWGAS VGGRSCEELT AVLTPPQLLG RRFNFFIQQK CGFRKAPRKV EPRRSDPGTS GEAYKRSALI PPVEETVFYP SPYPIRSLIK PLFFTVGFTG CAFGSAAIWQ YESLKSRVQS YFDGIKADWL DSIRPQKEGD FRKEINKWWN NLSDGQRTVT; Positive Samples: HepG2, MCF7; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein, Nucleus
This gene encodes a mitochondrial integral membrane protein. Following proteolytic processing of this protein, a small peptide is formed and translocated to the nucleus. This gene may be involved in signal transduction via regulated intramembrane proteolysis of membrane-tethered precursor proteins. Variation in this gene has been associated with increased risk for type 2 diabetes. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial intramembrane cleaving protease PARL; mitochondrial intramembrane-cleaving protease PARL; Presenilin-associated rhomboid-like protein, mitochondrial; rhomboid 7 homolog 1
Gene Aliases: PARL; PRO2207; PSARL; PSARL1; PSENIP2; RHBDS1
UniProt ID: (Human) Q9H300
Entrez Gene ID: (Human) 55486
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support