Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEPEFLYDL LQLPKGVEPP AEEELSKGGK KKYLPPTSRK DPKFEELQKV LMEWINATLL PEHIVVRSLE EDMFDGLILH HLFQRLAALK LEAEDIALTA TSQKHKLTVV LEAVNRSLQL EEWQAKWSVE SIFNKDLLST LHLLVALAKR FQPDLSLPTN VQVEVITIES TKSGLKSEKL VEQLTEYSTD KDEPPKDVFD ELFKLAPEKV NAVKEAIVNF VNQKLDRLGL SVQNLDTQFA DGVILLLLIG QLEGFFLHLK EFYLTPNSPA EMLHNVTLAL ELLKDEGLLS CPVSPEDIVN KDAKSTLRVL YGLFCKHTQK AHRDRTPHGA PN (1-331 aa encoded by BC034406)
Probably plays a role in the regulation of cell adhesion and cytoskeleton organization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gamma-parvin; parvin, gamma
Gene Aliases: AI413459; PARVG
UniProt ID: (Human) Q9HBI0, (Mouse) Q9ERD8
Entrez Gene ID: (Human) 64098, (Rat) 689069, (Mouse) 64099
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support