Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAWKSGGAS HSELIHNLRK NGIIKTDKVF EVMLATDRSH YAKCNPYMDS PQSIGFQATI SAPHMHAYAL ELLFDQLHEG AKALDVGSGS GILTACFARM VGCTGKVIGI DHIKELVDDS INNVRKDDPT LLSSGRVQLV VGDGRMGYAE EAPYDAIHVG AAAPVVPQAL IDQLKPGGRL ILPVGPAGGN QMLEQYDKLQ DGSIKMKPLM GVIYVPLTDK EKQWSRWK (1-227 aa encoded by BC008748)
Three classes of protein carboxyl methyltransferases, distinguished by their methyl-acceptor substrate specificity, have been found in prokaryotic and eukaryotic cells. The type II enzyme catalyzes the transfer of a methyl group from S-adenosyl-L-methionine to the free carboxyl groups of D-aspartyl and L-isoaspartyl residues. These methyl-accepting residues result from the spontaneous deamidation, isomerization, and racemization of normal L-aspartyl and L-asparaginyl residues and represent sites of covalent damage to aging proteins PCMT1 (EC 2.1.1.77) is a protein repair enzyme that initiates the conversion of abnormal D-aspartyl and L-isoaspartyl residues to the normal L-aspartyl form.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: L-isoaspartyl protein carboxyl methyltransferase; PIMT; protein carboxyl-o-methyltransferase; Protein L-isoaspartyl/D-aspartyl methyltransferase; Protein-beta-aspartate methyltransferase; Protein-L-isoaspartate(D-aspartate) O-methyltransferase
Gene Aliases: PCMT; PCMT1; PIMT
UniProt ID: (Pig) P80895, (Human) P22061
Entrez Gene ID: (Pig) 100502562, (Human) 5110
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support